Order Entry
Germany
ContactUsLinkComponent
3-Formyl-6-methoxypyridine
3-Formyl-6-methoxypyridine
Catalog # ASTA64881
Supplier:  ASTATECH
CAS Number:  
3-Formyl-6-methoxypyridine
Catalog # ASTA64881
Supplier:  ASTATECH
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :137.14 g/mol
CAS Number:65873-72-5
Formula:C₇H₇NO₂
MDL Number:MFCD02683446