


Tetrabutylammonium hydroxide 0.1 mol/l (0.1 N) in toluene 90% (v/v)/methanol 10% (v/v) Reag. Ph. Eur.
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:2052-49-5
:
Formula:C₁₆H₃₇NO
MDL Number:MFCD00009425
:259.48 g/mol
Storage Temperature:Refrigerator
EINECS:218-147-6
UN:1992
ADR:3,II
:GHS02
:GHS08
:GHS07
Frequently Bought Together