Order Entry
Germany
Orders LinkContactUsLinkComponent
Ethoxytrimethylsilane ≥97%

Ethoxytrimethylsilane ≥97%

Glass bottle for solids, 25 g
Catalog # ABCRAB111497
Flammable
Marketsource
Supplier:   abcr
Ethoxytrimethylsilane ≥97%
Catalog # ABCRAB111497
Supplier:  abcr
Marketsource
CAS Number:  1825-62-3
Ethoxytrimethylsilane ≥97%

Glass bottle for solids, 25 g

CAS Number:  1825-62-3
Price
184,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Trimethyl(ethoxy)silane
  • CAS Number:1825-62-3
  • Formula:C₅H₁₄OSi
  • MDL Number:MFCD00009069
  • Molecular Weight:118.25 g/mol
  • Boiling Pt:75…76 °C
  • Melting Pt:–83 °C
  • Density:0.757
  • Flash Pt:–4 °F
  • EINECS:217-370-6
  • UN:1993
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.