Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
8-Bromocaprylic acid
8-Bromocaprylic acid
8-Bromocaprylic acid
8-Bromocaprylic acid
FLUO001099-10G
:
FLUOROCHEM
:
8-Bromocaprylic acid
FLUO001099-10G
:
FLUOROCHEM
:
Pack type:
Glass bottle for solids
Pk:
10 g
Seq:
3236
:
:
223.11 g/mol
:
316 °C (1013 hPa)
:
316 °C (1013 hPa)
:
38.5 °C
:
38,5 °C
:
1,324 g/cm³ (20 °C)
:
17696-11-6
:
BrH₂C(CH₂)₆COOH
:
MFCD00004430