(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:14347-78-5
:
Formula:C₆H₁₂O₃
MDL Number:MFCD00003213
:132.16 g/mol
Merck Index:14,05213
Boiling Pt:78°C
Density:1,07 g/cm³ (20 °C)
Flash Pt:80°C
Storage Temperature:Refrigerator
EINECS:405-320-8
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
- Optical purity (GC):min. 98.0 ee%
- Specific rotation [a]20/D:-11.0 to -15.0 deg (neat)