Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Titanium(IV) propanolate
Titanium(IV) propanolate
Titanium(IV) propanolate
ABCRAKT885
:
abcr
:
Titanium(IV) propanolate
ABCRAKT885
:
abcr
:
Pack type:
Glass bottle for solids
Pk:
100 g
:
:
284.23 g/mol
:
137 °C (7 hPa)
:
1.033 g/cm³ (20 °C)
:
221-411-3
:
2413
:
3,III
:
3087-37-4
:
C₁₂H₂₈O₄Ti
:
MFCD00009365