(1S,2S)-(+)-1,2-Diaminocyclohexane
Catalog # FLUO012706-1G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:114.19 g/mol
Boiling Pt:187 °C (1013 hPa)
Melting Pt:40…43 °C
Storage Temperature:Refrigerator
UN:3259
ADR:8,II
CAS Number:21436-03-3
Formula:C₆H₁₄N₂
MDL Number:MFCD00062986