(3-(Dimethylamino)propyl)trimethoxysilane
Catalog # MSPPSID3546.94-10G
Supplier: MARKET SOURCE PART PROCESS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:207.35 g/mol
EINECS:219-786-3
CAS Number:2530-86-1
Formula:(H₃C)₂NCH₂CH₂CH₂Si(OCH₃)₃
MDL Number:MFCD00053674