
2-(2-(2-Chloroethoxy)ethoxy)ethanol
Catalog # ABCRAB139597.0025
Supplier: abcr
CAS Number:
Specifications
- Pk:25 g
Specifications
About this item
:
:168.62 g/mol
EINECS:225-984-0
:GHS07
CAS Number:5197-62-6
Formula:C₆H₁₃ClO₃
MDL Number:MFCD00002874