(S)-(+)-1-Benzyl-3-hydroxypiperidine
Catalog # FLUO011441-5G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:191.27 g/mol
CAS Number:91599-79-0
Formula:C₁₂H₁₇NO
MDL Number:MFCD03426359