Order Entry
Germany
ContactUsLinkComponent
1-Benzyl-4-hydroxypiperidine ≥97%
Irritant
1-Benzyl-4-hydroxypiperidine ≥97%
Catalog # ABCR10932
Supplier:  abcr
CAS Number:  
Irritant
1-Benzyl-4-hydroxypiperidine ≥97%
Catalog # ABCR10932
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

About this item

 :
 :191.27 g/mol
Boiling Pt:121…123 °C (0.7 mmHg)
Melting Pt:60…64 °C
EINECS:225-226-9
 :GHS07
Storage Temperature:Ambient
CAS Number:4727-72-4
Formula:C₁₂H₁₇NO
MDL Number:MFCD00006503