
Ethyl-2-methylnicotinate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1721-26-2
:
Formula:C₉H₁₁NO₂
MDL Number:MFCD00006336
:165.19 g/mol
Boiling Pt:107…108 °C (13 mmHg)
Density:1.070
Flash Pt:102 °C (215 °F)
EINECS:217-013-4
:GHS07
Storage Temperature:Ambient