Fluoride standard solution, 1 mg/L F- in Water (from NaF) according to DIN 38405-4:1985, DEV D4
Supplier: Bernd Kraft
Certificates
Fluoride standard solution, 1 mg/L F- in Water (from NaF) according to DIN 38405-4:1985, DEV D4
Supplier: Bernd Kraft
About this item
Frequently Bought Together
Disinfectant solutions, Bacillol AF
Supplier: BODE CHEMIE
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Detergents, liquid, for automated cleaning of glassware, neodisher® FLA
Supplier: Dr. Weigert
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Neutralising agent for the automated cleaning of glassware, neodisher® Z
Supplier: Dr. Weigert
Buffers, concentrated solutions, Titrisol®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
VWR® Aquaresist Water Bath Protective Agent
Supplier: VWR Chemicals
pH electrode, SE 100 N
Supplier: KNICK
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals