4-Methyl-3-(trifluoromethyl)benzoic acid
Catalog # MOLEM56718436
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:204.15 g/mol
CAS Number:261952-01-6
Formula:C₉H₇F₃O₂
MDL Number:MFCD01631592