Order Entry
Germany
ContactUsLinkComponent
 
2-Chloro-N-(2-ethyl-6-methylphenyl)acetamide
Catalog # EHERC13348010
Supplier:  EHRENSTORFER
CAS Number:  
2-Chloro-N-(2-ethyl-6-methylphenyl)acetamide
Catalog # EHERC13348010
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :211.6880 g/mol
CAS Number:32428-71-0
Formula:C₁₁H₁₄ClNO
MDL Number:MFCD00267498