Order Entry
Germany
ContactUsLinkComponent
Arsenic standard solution, 10 mg/L as in dil. nitric acid standard for ICP-MS
Arsenic standard solution, 10 mg/L as in dil. nitric acid standard for ICP-MS
Catalog # 89189.180
Supplier:  CPACHEM
CAS Number:  
Arsenic standard solution, 10 mg/L as in dil. nitric acid standard for ICP-MS
Catalog # 89189.180
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    2-5% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Arsenic
  • Symbol:
    As

Specifications

About this item