3-Amino-3-(2-methylphenyl)propionic acid
Marketsource
Supplier: BIONET RESEARCH
Jump to:
About this item
Synonyms: 3-Amino-3-(o-tolyl)propanoic acid
- CAS Number:68208-16-2
- Formula:C₁₀H₁₃NO₂
- MDL Number:MFCD01871311
- Molecular Weight:179.2200 g/mol
2 Options Available Below
Product Details & Documents
Synonyms: 3-Amino-3-(o-tolyl)propanoic acid
- CAS Number:68208-16-2
- Formula:C₁₀H₁₃NO₂
- MDL Number:MFCD01871311
- Molecular Weight:179.2200 g/mol
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource



