Order Entry
Germany
Orders LinkContactUsLinkComponent
3-Bromo-p-anisidine

3-Bromo-p-anisidine

Glass bottle for solids, 25 g
Catalog # FLUO038608-25G
Marketsource
Supplier:   FLUOROCHEM
3-Bromo-p-anisidine
Catalog # FLUO038608-25G
Supplier:  FLUOROCHEM
Marketsource
CAS Number:  19056-41-8
3-Bromo-p-anisidine

Glass bottle for solids, 25 g

CAS Number:  19056-41-8
Price
129,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  3-Bromo-4-methoxyaniline
  • CAS Number:19056-41-8
  • Formula:C₇H₈BrNO
  • MDL Number:MFCD04094339
  • Molecular Weight:202.0485 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.