


Zinc sulphate 0.3 mol/l (0.3 N) in aqueous solution, Sigma-Aldrich®
Catalog # SIALZ2876-500ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:500 mL
- Seq:2725
Specifications
About this item
:
:161.45 g/mol
Storage Temperature:Ambient
EINECS:231-793-3
UN:3082
ADR:9,III
:GHS05
:GHS07
:GHS09
CAS Number:7733-02-0
Formula:O₄SZn
MDL Number:MFCD00011302