Order Entry
Germany
ContactUsLinkComponent
Magnesium hexafluorosilicate hexahydrate
Magnesium hexafluorosilicate hexahydrate
Catalog # HONE01415.9025
CAS Number:  
Magnesium hexafluorosilicate hexahydrate
Catalog # HONE01415.9025
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

About this item

 :
 :274.47 g/mol
EINECS:241-022-2
UN:2853
ADR:6.1,III
CAS Number:18972-56-0
Formula:F₆MgSi·6H₂O
MDL Number:MFCD00150250