Benzaldehyde azine
Catalog # FLUO018494-10G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:208.27 g/mol
EINECS:209-627-6
CAS Number:588-68-1
Formula:C₁₄H₁₂N₂
MDL Number:MFCD00016876