5-Methyl-4-phenyl-o-anisidine
Catalog # FLUO018853-1G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
CAS Number:206761-76-4
Formula:C₁₄H₁₅NO
MDL Number:MFCD00270117