Tolbutamide
Catalog # EHERC17589900
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:270.35 g/mol
EINECS:200-594-3
CAS Number:64-77-7
Formula:C₁₂H₁₈N₂O₃S
MDL Number:MFCD00027169