Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Amino-4-(4-fluorophenyl)thiazole
2-Amino-4-(4-fluorophenyl)thiazole
2-Amino-4-(4-fluorophenyl)thiazole
2-Amino-4-(4-fluorophenyl)thiazole
Supplier:
BIONET RESEARCH
Certificates
Expand image
2-Amino-4-(4-fluorophenyl)thiazole
Supplier:
BIONET RESEARCH
About this item
CAS Number:
77815-14-6
:
Formula:
C₉H₇FN₂S
MDL Number:
MFCD01086598
:
194.2287 g/mol