Order Entry
Germany
Orders LinkContactUsLinkComponent
Methyl-2-formylbenzoate

Methyl-2-formylbenzoate

Marketsource
Supplier:   Thermo Scientific
Methyl-2-formylbenzoate
Supplier:  Thermo Scientific
Marketsource
Certificates
Methyl-2-formylbenzoate

3 Options Available Below

Product Details & Documents
Synonyms:  Methyl 2-formylbenzoate
  • CAS Number:4122-56-9
  • Formula:C₉H₈O₃
  • MDL Number:MFCD00204231
  • Molecular Weight:164.1580 g/mol
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.