Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-N-tert-butylbenzenesulphonamide
2-Bromo-N-tert-butylbenzenesulphonamide
2-Bromo-N-tert-butylbenzenesulphonamide
2-Bromo-N-tert-butylbenzenesulphonamide
Supplier:
COMBI-BLOCKS
Certificates
Expand image
2-Bromo-N-tert-butylbenzenesulphonamide
Supplier:
COMBI-BLOCKS
About this item
CAS Number:
138733-50-3
:
Formula:
C₁₀H₁₄BrNO₂S
MDL Number:
MFCD07710752