Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Amino-3-(2-chlorophenyl)propionic acid
3-Amino-3-(2-chlorophenyl)propionic acid
3-Amino-3-(2-chlorophenyl)propionic acid
3-Amino-3-(2-chlorophenyl)propionic acid
Supplier:
BIONET RESEARCH
Certificates
Expand image
3-Amino-3-(2-chlorophenyl)propionic acid
Supplier:
BIONET RESEARCH
About this item
CAS Number:
68208-20-8
:
Formula:
C₉H₁₀ClNO₂
MDL Number:
MFCD01871299
:
199.64 g/mol