
Methyl-3-amino-5,6-dichloropyrazine-2-carboxylate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1458-18-0
:
Formula:C₆H₅Cl₂N₃O₂
MDL Number:MFCD00010431
:222.03 g/mol
Melting Pt:230…233 °C
EINECS:215-947-7
:GHS07
Storage Temperature:Ambient