Order Entry
Germany
ContactUsLinkComponent
Methyl-5-chloropyrazine-2-carboxylate, Sigma-Aldrich®
Methyl-5-chloropyrazine-2-carboxylate, Sigma-Aldrich®
Catalog # SIAL688584-5G
Supplier:  Merck
CAS Number:  
Methyl-5-chloropyrazine-2-carboxylate, Sigma-Aldrich®
Catalog # SIAL688584-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :172.57 g/mol
CAS Number:33332-25-1
Formula:C₆H₅ClN₂O₂
MDL Number:MFCD01632102