Order Entry
Germany
Orders LinkContactUsLinkComponent
Chromone-3-carboxaldehyde ≥97%

Chromone-3-carboxaldehyde ≥97%

Chromone-3-carboxaldehyde ≥97%
Certificates
Chromone-3-carboxaldehyde ≥97%
Chromone-3-carboxaldehyde ≥97%

3 Options Available Below

Product Details & Documents

Purity: 97% CAS No.: 17422-74-1 Molecular Formula: C10H6O3 Formula Weight: 174.16 Storage Temperature: 2°C to 8°C Melting Point: 150-154° Beilstein Reference: 472849 MDL No.: MFCD00014667

Synonyms:  4-Oxo-4H-chromene-3-carbaldehyde
  • CAS Number:17422-74-1
  • Formula:C₁₀H₆O₃
  • MDL Number:MFCD00014667
  • Molecular Weight:174.16 g/mol
  • Melting Pt:150…154 °C
  • EINECS:241-451-5
Product Family Options

Product Information