Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dimethylmethoxysilane
Dimethylmethoxysilane
Dimethylmethoxysilane
Dimethylmethoxysilane
Supplier:
CHEMPUR SILANE
Certificates
Expand image
Dimethylmethoxysilane
Supplier:
CHEMPUR SILANE
About this item
CAS Number:
18033-75-5
:
Formula:
C₃H₁₀OSi