1-Hydroxy-4-(p-toluidino)-9,10-anthraquinone
Catalog # EHERC16971330
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:329.3500 g/mol
EINECS:201-353-5
CAS Number:81-48-1
Formula:C₂₁H₁₅NO₃
MDL Number:MFCD00019148