Order Entry
Germany
ContactUsLinkComponent
Quinizarin green 95% (dye content), Sigma-Aldrich®
Irritant
Quinizarin green 95% (dye content), Sigma-Aldrich®
Catalog # 211982-50G
CAS Number:  
undefined
Irritant
Quinizarin green 95% (dye content), Sigma-Aldrich®
Catalog # 211982-50G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • Appearance (Form):
    Powder
  • Infrared spectrum:
    Conforms to Structure
  • C=0.01 G/L Acetone:
    624 - 644 nm
  • Extinction Coefficient:
    ≥15000
  • Wavelength:
    591 - 611 nm
  • Extinction Coefficient, In Acetone:
    ≥6000
  • Carbon:
    76.3 - 84.4 %
  • Nitrogen:
    6.3 - 7.0 %
  • TLC:
    Consistent with Past Lots
  • Solubility (Turbidity):
    Turbid
  • Solubility (Color) c = 1%, Pyridine:
    Dark Blue to Very Dark Blue and Dark, Blue-Green to Very Dark Blue-Green

Specification Test Results

About this item

 :
 :418.496 g/mol
Melting Pt:220...221 °C (lit.)
 :M-Clarity™ quality level = MQ200
EINECS:204-909-5
 :GHS07
CAS Number:128-80-3
Formula:C₂₈H₂₂N₂O₂
MDL Number:MFCD00001195