2-Chloro-4-phenylthiazol
Catalog # FLUO024114-250MG
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:195.668 g/mol
CAS Number:1826-23-9
Formula:C₉H₆ClNS
MDL Number:MFCD00956371