Order Entry
Germany
ContactUsLinkComponent
4-Bromo-1,8-naphthalic anhydride, Sigma-Aldrich®
4-Bromo-1,8-naphthalic anhydride, Sigma-Aldrich®
Catalog # SIAL324248-5G
Supplier:  Merck
CAS Number:  
4-Bromo-1,8-naphthalic anhydride, Sigma-Aldrich®
Catalog # SIAL324248-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :277.0703 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:201-382-3
CAS Number:81-86-7
Formula:C₁₂H₅BrO₃
MDL Number:MFCD00006927