HBTU(O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate) 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:94790-37-1
:
Formula:C₁₁H₁₆F₆N₅OP
MDL Number:MFCD00075445
:379.2 g/mol
Melting Pt:200 °C (dec.)
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Solubility:(10 % in DMF) Clear colorless
- HPLC:≥97.5 %