HBTU(O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate)
Catalog # ABCRAB128869.0025
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
About this item
:
:379.2 g/mol
CAS Number:94790-37-1
Formula:C₁₁H₁₆F₆N₅OP
MDL Number:MFCD00075445