Order Entry
Germany
ContactUsLinkComponent
TATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate)
TATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate)
Catalog # MOLE67943083-5G
Supplier:  Molekula
CAS Number:  
TATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate)
Catalog # MOLE67943083-5G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :322.1 g/mol
CAS Number:873798-09-5
Formula:C₁₀H₁₅BF₄N₆O
MDL Number:MFCD08064301