Order Entry
Germany
Orders LinkContactUsLinkComponent
Estriol

Estriol

Glass bottle for solids, 100 mg
Catalog # PROOMM0130.00
Estriol
Catalog # PROOMM0130.00
CAS Number:  50-27-1
Estriol

Glass bottle for solids, 100 mg

CAS Number:  50-27-1
Price
1 290,00 €
Log in for availability
1 * 100 mg
Product Details & Documents
  • CAS Number:50-27-1
  • Formula:C₁₈H₂₄O₃
  • MDL Number:MFCD00003691
  • EINECS:200-022-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.