Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(2-Thienyl)-1,3-thiazolidine-4-carboxylic acid
2-(2-Thienyl)-1,3-thiazolidine-4-carboxylic acid
2-(2-Thienyl)-1,3-thiazolidine-4-carboxylic acid
2-(2-Thienyl)-1,3-thiazolidine-4-carboxylic acid
Supplier:
FLUOROCHEM
Certificates
Expand image
2-(2-Thienyl)-1,3-thiazolidine-4-carboxylic acid
Supplier:
FLUOROCHEM
About this item
CAS Number:
32451-19-7
:
Formula:
C₈H₉NO₂S₂
MDL Number:
MFCD00022489
EINECS:
251-055-4