Order Entry
Germany
ContactUsLinkComponent
 
 
 
2-Mercaptopyridine-N-oxide sodium salt 40% (w/w) in aqueous solution
 
undefined
 
 
2-Mercaptopyridine-N-oxide sodium salt 40% (w/w) in aqueous solution

 

  • Appearance:
    Yellow to amber Solution
  • pH:
    8.5 to 10.5 (10 % at 20°C)
  • Titration with HClO4:
    40 to 42 %