Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')terbium(III) ≥99% (99.9%-Tb)
Catalog # STRE65-8000-1G
Supplier: Strem Chemicals, Inc.
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:708.727 g/mol
Boiling Pt:275 °C (dec.)
Melting Pt:155…156 °C
CAS Number:15492-51-0
Formula:C₃₃H₅₇O₆Tb
MDL Number:MFCD00064752