Order Entry
Germany
ContactUsLinkComponent
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')aluminium ≥99% (99.9%-Al)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')aluminium ≥99% (99.9%-Al)
Catalog # STRE13-5000-1G
CAS Number:  
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')aluminium ≥99% (99.9%-Al)
Catalog # STRE13-5000-1G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :576.78 g/mol
Boiling Pt:>400 °C/ 150 °C (0.01 mmHg) (dec./ subl.)
Boiling Pt:>400 °C/ 150 °C (0,01 mmHg) (dec./ subl.)
Melting Pt:255…258 °C
CAS Number:14319-08-5
Formula:C₃₃H₅₇AlO₆
MDL Number:MFCD00058918