Order Entry
Germany
ContactUsLinkComponent
Gel filtration media, Sephadex™ G-75
Gel filtration media, Sephadex™ G-75
Supplier:  Cytiva
Certificates
undefined
Gel filtration media, Sephadex™ G-75
Supplier:  Cytiva

Specifications

  • pH range:
    2 - 10
  • Matrix:
    Cross-linked dextran
  • Fractionation range:
    1 × 103 - 5 × 104 dextrans, 3 × 103 - 8 × 104 globular proteins
  • Storage temperature:
    4 - 30 °C

Specifications

Related Information