Order Entry
Germany
ContactUsLinkComponent
Hydrometer for buttermilk
Hydrometer for buttermilk
Catalog # GERB6640
Supplier:  GERBER FUNKE
undefined
Hydrometer for buttermilk
Catalog # GERB6640
Supplier:  GERBER FUNKE

Specifications

  • Description:
    Hydrometer for buttermilk
  • Length:
    240 mm
  • Calibration temperature:
    20 °C
  • Division:
    0,0002 g/ml
  • Pk:
    1
  • Range:
    1,014 - 1,030 g/ml

Specifications

About this item

Hydrometer for milk analysis, especially butter milk serum.

Certifications: DIN 10293