Disposable earplugs, MelLows®
Supplier: Moldex
Certificates
About this item
Earplugs made from soft PU-foam for protection against lower noise levels.
SNR: 22 dB
Certifications: EN 352-2:2002
Ordering information: Also available in the hygienic dispenser system MoldexStation.
Packaging: In pairs.
Specifications
- Catalog No:
- MOLD7625
- MOLD7600
- MOLD7650
- Environmentally Preferable:
- Safer Human and Environmental Health