Hydrogen peroxide 30% stabilised, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:7722-84-1
:
Formula:H₂O₂
MDL Number:MFCD00011333
Boiling Pt:107 °C (1013 hPa)
Melting Pt:? 0 °C
Density:1.11 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-765-0
UN:2014
ADR:5.1 (8),II
:34.01 g/mol
:GHS03
:GHS05
:GHS06
:GHS08
Specification Test Results
- Meets ACS Reagent Chemical Requirements:30.0 - 32.0 %
- For Laboratory,Research,or Manufacturing Use:Max. 10
- Assay (H₂O₂):30.0 - 32.0 %
- Color (APHA):≤10
- Residue after Evaporation:≤0.0020 %
- Titrable Acid (meq/g):≤0.0006
- Chloride (Cl):≤2 ppm
- Nitrate (NO₃):≤2 ppm
- Phosphate (PO₄):≤2 ppm
- ACS - Sulfate (SO₄):≤5 ppm
- Ammonium (NH₄):≤5 ppm
- Trace Impurities - Copper (Cu):≤50 ppb
- Trace Impurities - Iron (Fe):≤100.0 ppb
- Heavy Metals (as Pb):≤1000.0 ppb
- Trace Impurities - Lead (Pb):≤100 ppb
- Trace Impurities - Nickel (Ni):≤50 ppb