IgG purification kits, Pearl™
Supplier: G-Biosciences
Certificates
About this item
The Pearl™ IgG purification kits are designed for the one step purification of IgG antibodies from serum. The supplied Pearl™ IgG purification resin binds the high abundant, non-IgG proteins (i.e. albumin) and allows the IgG molecules to pass through in a physiological buffer. The purified IgG molecules can be stored or used in further downstream applications without further clean-up, such as ammonium sulphate precipitation.
- One step purification
- Purification in <15 minutes
- Faster purification compared to Protein A or Protein G resins
- >90% recovery with >80% purity
The Pearl™ IgG purification kits are available in two formats - a spin format that is ideal for the rapid, small scale purification of IgG or a larger format kit for the isolation of IgG from ~100 ml serum (~200 mg 1gG).
Delivery: The spin format kit is supplied with 3 ml Pearl™ IgG purification resin, IgG isolation buffer and 20 spin columns. The large format kit contains 25 ml Pearl™ IgG purification resin and IgG isolation buffer.