SparkPlugs® Disposable Earplugs
: Moldex
SparkPlugs® earplugs are made of a soft foam which becomes firmer when rolled for insertion.
- Tapered shape fits easily in the ear canal
- Seals gently and snugly without pressure
- Appealing colours
SNR: 35 dB
: EN 352-2:2002
: Also available in the hygienic MoldexStation dispenser system.
: * One size fits all.