Graduated pipettes, SILBERBRAND ETERNA, type 2, Class B, total delivery
Supplier: Brand
Certificates
About this item
AR-Glas®.
- Calibrated to deliver (TD, Ex)
- Nominal volume at the top
DIN EN ISO 835
Length: 360 mm ±10 mm except for 25 ml pipettes: 450 mm ±10 mm.
Certifications: DIN EN ISO 835
Caution: * Suitable for cotton plugs